Uridine, 99%
Uridine, 99%
Uridine, 99%
Thermo Scientific Chemicals

Uridine, 99%

Uridine, 99%, C9H12N2O6, CAS Number-58-96-8, uracil, 1-beta-d-ribofuranosyl, uracil riboside, d-uridine, beta-uridine, 1-beta-d-ribofuranosyluracil, uridin, unii-whi7hq7h85, urd, b-uridine, uridine, 100g, 754904, 200-407-5, 244.2, DRTQHJPVMGBUCF-DHRBYNEYSA-N | CAS: 58-96-8 | C9H12N2O6 | 244.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15227.14
also known as A15227-14
Price (EUR)
117,65
Online Exclusive
130,00
Save 12,35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
117,65
Online Exclusive
130,00
Save 12,35 (10%)
Each
Chemical Identifiers
CAS58-96-8
IUPAC Name1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H12N2O6
InChI KeyDRTQHJPVMGBUCF-UHFFFAOYNA-N
SMILESOCC1OC(C(O)C1O)N1C=CC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)white to pale cream
Assay (HPLC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)164-172?C
Optical Rotation+9.0 ? 1.5? (c=2, water)
View more
Uridine plays a vital role in the glycolysis pathway of galactose. It is used as a precursor in the production of CDP-choline. It is an important nutrient and widely used as a dietary supplement. It improves brain cholinergic functions and hepatic mitochondrial function in certain liver toxins. It plays a major role in pain physiology and brain energy utilization to maintain ATP production under restricted oxygen conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Uridine plays a vital role in the glycolysis pathway of galactose. It is used as a precursor in the production of CDP-choline. It is an important nutrient and widely used as a dietary supplement. It improves brain cholinergic functions and hepatic mitochondrial function in certain liver toxins. It plays a major role in pain physiology and brain energy utilization to maintain ATP production under restricted oxygen conditions.

Solubility
Soluble in water, dimethylsulfoxide, and methanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Heuberger, B. D.; Pal, A.; Del Frate, F.; Topkar, V. V.; Szostak, J. W. Replacing Uridine with 2-Thiouridine Enhances the Rate and Fidelity of Nonenzymatic RNA Primer Extension. J. Am. Chem. Soc. 2015, 137 (7), 2769-2775.
  2. L Ipata, P.; Pesi, R. Metabolic Regulation of Uridine in the Brain. Curr Metabolomics 2015, 3 (1), 4-9.