3-Amino-5-mercapto-1,2,4-triazole, 97+%
3-Amino-5-mercapto-1,2,4-triazole, 97+%
3-Amino-5-mercapto-1,2,4-triazole, 97+%
Thermo Scientific Chemicals

3-Amino-5-mercapto-1,2,4-triazole, 97+%

CAS: 16691-43-3 | C2H4N4S | 116.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15242.06
also known as A15242-06
Price (EUR)
29,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
29,30
Each
Chemical Identifiers
CAS16691-43-3
IUPAC Name5-amino-2,3-dihydro-1H-1,2,4-triazole-3-thione
Molecular FormulaC2H4N4S
InChI KeyWZUUZPAYWFIBDF-UHFFFAOYSA-N
SMILESNC1=NC(=S)NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
Assay (HPLC)≥97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only