4-Chlorocinnamic acid, predominantly trans, 99%
4-Chlorocinnamic acid, predominantly trans, 99%
4-Chlorocinnamic acid, predominantly trans, 99%
4-Chlorocinnamic acid, predominantly trans, 99%
Thermo Scientific Chemicals

4-Chlorocinnamic acid, predominantly trans, 99%

CAS: 1615-02-7 | C9H7ClO2 | 182.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A15304.22
also known as A15304-22
Price (EUR)
132,60
Online Exclusive
156,00
Save 23,40 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
132,60
Online Exclusive
156,00
Save 23,40 (15%)
Each
Add to cart
Chemical Identifiers
CAS1615-02-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)246.0-252.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only