Creatine phosphate disodium salt tetrahydrate, 98+%
Creatine phosphate disodium salt tetrahydrate, 98+%
Creatine phosphate disodium salt tetrahydrate, 98+%
Thermo Scientific Chemicals

Creatine phosphate disodium salt tetrahydrate, 98+%

CAS: 71519-72-7 | C4H8N3Na2O5P·4H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A15362.06
also known as A15362-06
Price (EUR)
95,65
Online exclusive
106,00
Save 10,35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
95,65
Online exclusive
106,00
Save 10,35 (10%)
Each
Chemical Identifiers
CAS71519-72-7
IUPAC Namedisodium tetrahydrate ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
Molecular FormulaC4H16N3Na2O9P
InChI KeyHUWYWJSJJDCZRQ-UHFFFAOYSA-L
SMILESO.O.O.O.[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystalline powder
Impurity contentCreatine: ≤0.6%
Assay from Supplier's CofA≥98.0% (HPLC - Dry wt. basis)
Water Content (Karl Fischer Titration)≥20.0 to ≤25.5% (3.54-4.85 waters)
Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.

Solubility
Soluble in water.0.1 g/mL.

Notes
Incompatible with Strong bases. strong acids. Protect from Direct sunlight.
RUO – Research Use Only
A. H. Ennor and L. A. Stocken. The preparation of sodium phosphocreatine.Biochem J.1948, 43 (2), 190-191.