Thermo Scientific Chemicals

1,1,4,7,7-Pentamethyldiethylenetriamine, 98%, Thermo Scientific Chemicals

Catalog number: A15433.22
100 g, Each
Thermo Scientific Chemicals

1,1,4,7,7-Pentamethyldiethylenetriamine, 98%, Thermo Scientific Chemicals

Catalog number: A15433.22
100 g, Each
Quantity
Catalog number: A15433.22
also known as A15433-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
3030-47-5
IUPAC Name
(2-{[2-(dimethylamino)ethyl](methyl)amino}ethyl)dimethylamine
Molecular Formula
C9H23N3
InChI Key
UKODFQOELJFMII-UHFFFAOYSA-N
SMILES
CN(C)CCN(C)CCN(C)C
Refractive Index
1.4400-1.4450 @ 20?C
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Form
Liquid

Description

Used as ligand in NMR study of complexation/aggregation with neopentyllithium1 and in the study of ligand effects on the selectivity of CO2 incorporation into α,ω-diynes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as ligand in NMR study of complexation/aggregation with neopentyllithium1 and in the study of ligand effects on the selectivity of CO2 incorporation into α,ω-diynes

Solubility
Miscible in water.

Notes
Air & Moisture Sensitive. Store under inert gas. Incompatible with strong oxidizing agents, strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 3
    Acute Inhalation Toxicity - Vapors
    Category 3
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Chronic aquatic toxicity
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 - Harmful if swallowed

    H311 + H331 - Toxic in contact with skin or if inhaled

    H314 - Causes severe skin burns and eye damage

    H412 - Harmful to aquatic life with long lasting effects

    EU Specific Hazard Statements

    Combustible liquid

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician