2-Ethylfuran, 98+%
2-Ethylfuran, 98+%
2-Ethylfuran, 98+%
Thermo Scientific Chemicals

2-Ethylfuran, 98+%

CAS: 3208-16-0 | C6H8O | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15520.22
also known as A15520-22
Price (EUR)
397,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
397,00
Each
Chemical Identifiers
CAS3208-16-0
IUPAC Name2-ethylfuran
Molecular FormulaC6H8O
InChI KeyHLPIHRDZBHXTFJ-UHFFFAOYSA-N
SMILESCCC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Transition Temperature1.4380-1.4420 @ 20?C
Assay (GC)>97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only