4'-(Trifluoromethyl)acetophenone, 98+%
4'-(Trifluoromethyl)acetophenone, 98+%
4'-(Trifluoromethyl)acetophenone, 98+%
Thermo Scientific Chemicals

4'-(Trifluoromethyl)acetophenone, 98+%

CAS: 709-63-7 | C9H7F3O | 188.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15529.06
also known as A15529-06
Price (EUR)
55,50
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
55,50
Each
Chemical Identifiers
CAS709-63-7
IUPAC Name1-[4-(trifluoromethyl)phenyl]ethan-1-one
Molecular FormulaC9H7F3O
InChI KeyHHAISVSEJFEWBZ-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder or fused solid
Identification (FTIR)Conforms
Appearance (Color)White to pale cream or pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only