3-(4-Morpholinyl)phenol, 98%
3-(4-Morpholinyl)phenol, 98%
3-(4-Morpholinyl)phenol, 98%
Thermo Scientific Chemicals

3-(4-Morpholinyl)phenol, 98%

CAS: 27292-49-5 | C10H13NO2 | 179.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A15707.14
also known as A15707-14
Price (EUR)
485,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
485,00
Each
Chemical Identifiers
CAS27292-49-5
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to cream to pale brown
Melting Point (clear melt)127.5-133.5?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only