Choline chloride, 98+%
Choline chloride, 98+%
Choline chloride, 98+%
Thermo Scientific Chemicals

Choline chloride, 98+%

CAS: 67-48-1 | C5H14ClNO | 139.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A15828.0E
also known as A15828-0E
Price (EUR)
209,65
Online exclusive
233,00
Save 23,35 (10%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (EUR)
209,65
Online exclusive
233,00
Save 23,35 (10%)
Each
Chemical Identifiers
CAS67-48-1
IUPAC Name(2-hydroxyethyl)trimethylazanium chloride
Molecular FormulaC5H14ClNO
InChI KeySGMZJAMFUVOLNK-UHFFFAOYSA-M
SMILES[Cl-].C[N+](C)(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or needles or adhering crystals
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤2%
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.

Solubility
Soluble in water, ethanol, methanol and alcohol.

Notes
Incompatible with strong oxidizing agents, strong bases, strong acids, alkalies and alloys.
RUO – Research Use Only

General References:

  1. Sun, S.; Niu, Y.; Xu, Q.; Sun, Z.; Wei, X. Efficient SO2 Absorptions by Four Kinds of Deep Eutectic Solvents Based on Choline Chloride. Ind. Eng. Chem. Res. 2015, 54 (33), 8019-8024.
  2. Das, A.; Biswas, R. Dynamic Solvent Control of a Reaction in Ionic Deep Eutectic Solvents: Time-Resolved Fluorescence Measurements of Reactive and Nonreactive Dynamics in (Choline Chloride + Urea) Melts. J. Phys. Chem. B 2015, 119 (31), 10102-10113.