1-Decanethiol, 96%
1-Decanethiol, 96%
1-Decanethiol, 96%
Thermo Scientific Chemicals

1-Decanethiol, 96%

CAS: 143-10-2 | C10H22S | 174.346 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Catalog number A15913.AE
also known as A15913-AE
Price (EUR)
110,00
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
110,00
Each
Chemical Identifiers
CAS143-10-2
IUPAC Namedecane-1-thiol
Molecular FormulaC10H22S
InChI KeyVTXVGVNLYGSIAR-UHFFFAOYSA-N
SMILESCCCCCCCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
CommentApparent color may differ on changing container
Appearance (Color)Clear colorless to pale blue or pale green
Refractive Index1.4540-1.4590 @ 20?C
FormLiquid
Assay (GC)≥95.0%
1-Decanethiol is used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for field-effect transistors (FETs). It is also used in the study of self-assembled monolayers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decanethiol is used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for field-effect transistors (FETs). It is also used in the study of self-assembled monolayers.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. It is sensitive to air. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. M. Elena Pérez-Bernal.; Ricardo Ruano-Casero.; Thomas J. Pinnavaia. Catalytic autoxidation of 1-decanethiol by cobalt(II) phthalocyaninetetrasulfonate intercalated in a layered double hydroxide. Catalysis Letters. 1991, 11 (1), 55-61.
  2. N. Isa.; K. D. Gibson.; T. Yan.; W. Hase.; S. J. Sibener. Experimental and simulation study of neon collision dynamics with a 1-decanethiol monolayer. J. Chem. Phys. 2000, 120 2417.