4-Bromobenzotrifluoride, 99%
4-Bromobenzotrifluoride, 99%
4-Bromobenzotrifluoride, 99%
Thermo Scientific Chemicals

4-Bromobenzotrifluoride, 99%

CAS: 402-43-7 | C7H4BrF3 | 225.008 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15942.22
also known as A15942-22
Price (EUR)
447,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
447,00
Each
Chemical Identifiers
CAS402-43-7
IUPAC Name1-bromo-4-(trifluoromethyl)benzene
Molecular FormulaC7H4BrF3
InChI KeyXLQSXGGDTHANLN-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Refractive Index1.4715-1.4745 @ 20?C
FormCrystals or powder or crystalline powder or clear liquid as melt
Identification (FTIR)Conforms
Appearance (Color)Colorless to white to pale yellow
4-Bromobenzotrifluoride is used to study the catalytic activity of palladacycle using Suzuki-Miyaura reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzotrifluoride is used to study the catalytic activity of palladacycle using Suzuki-Miyaura reaction.

Solubility
Not miscible or difficult to mix.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which areopened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Woen Susanto.; Chi-Yuan Chu.; Wei Jie Ang,.; Tzyy-Chao Chou.; Lee-Chiang Lo.; Yulin Lam. Development of a fluorous, oxime-based palladacycle for microwave-promoted carbon-carbon coupling reactions in aqueous media. Green Chem. 2012, 14 77-80 .
  2. Mark Sundermeier Dr., Alexander Zapf Dr.; Matthias Beller Prof. Dr. A Convenient Procedure for the Palladium-Catalyzed Cyanation of Aryl Halides. Angewandte Chemie International Edition. 2003, 42 (14), 1661-1664.