4-Bromoisoquinoline, 98%
4-Bromoisoquinoline, 98%
4-Bromoisoquinoline, 98%
Thermo Scientific Chemicals

4-Bromoisoquinoline, 98%

CAS: 1532-97-4 | C9H6BrN | 208.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15951.22
also known as A15951-22
Price (EUR)
555,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
555,00
Each
Chemical Identifiers
CAS1532-97-4
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid or clear liquid as melt
Appearance (Color)Colorless to cream to pale yellow
Melting Point (clear melt)38.0-45.0?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only