6-Methoxy-1-tetralone, 99%
6-Methoxy-1-tetralone, 99%
6-Methoxy-1-tetralone, 99%
Thermo Scientific Chemicals

6-Methoxy-1-tetralone, 99%

CAS: 1078-19-9 | C11H12O2 | 176.215 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15967.06
also known as A15967-06
Price (EUR)
22,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
22,30
Each
Chemical Identifiers
CAS1078-19-9
IUPAC Name6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one
Molecular FormulaC11H12O2
InChI KeyMNALUTYMBUBKNX-UHFFFAOYSA-N
SMILESCOC1=CC=C2C(=O)CCCC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)74.0-81.0?C
Appearance (Color)Cream to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only