1-Octanol, 99%
1-Octanol, 99%
1-Octanol, 99%
Thermo Scientific Chemicals

1-Octanol, 99%

CAS: 111-87-5 | C8H18O | 130.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
500 mL
2500 mL
Catalog number A15977.0D
Price (EUR)
243,65
キャンペーン価格
271,00
Save 27,35 (10%)
Each
Quantity:
10,000 mL
Request bulk or custom format
Price (EUR)
243,65
キャンペーン価格
271,00
Save 27,35 (10%)
Each
Chemical Identifiers
CAS111-87-5
IUPAC Nameoctan-1-ol
Molecular FormulaC8H18O
InChI KeyKBPLFHHGFOOTCA-UHFFFAOYSA-N
SMILESCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Refractive Index1.4280-1.4310 @ 20?C
FormLiquid
Assay (GC)≥98.5%
1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.

Solubility
Miscible with ethanol, ether and propylene glycol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Akhtar, M. a.; Dandapani, H.; Thiel, K.; Jones, P. R. Microbial production of 1-octanol: A naturally excreted biofuel with diesel-like properties. Metab. Eng. Commun. 2015, 2, 1-5.
  2. Mortensen, P. M.; de Carvalho, H. W. P.; Grunwaldt, J. D.; Jensen, P. A.; Jensen, A. D. Activity and stability of Mo2C/ZrO2 as catalyst for hydrodeoxygenation of mixtures of phenol and 1-octanol. J. Catal. 2015, 328, 208-215.