1,3,5-Trifluorobenzene, 98+%
1,3,5-Trifluorobenzene, 98+%
1,3,5-Trifluorobenzene, 98+%
Thermo Scientific Chemicals

1,3,5-Trifluorobenzene, 98+%

CAS: 372-38-3 | C6H3F3 | 132.085 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15984.06
also known as A15984-06
Price (EUR)
69,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
69,20
Each
Chemical Identifiers
CAS372-38-3
IUPAC Name1,3,5-trifluorobenzene
Molecular FormulaC6H3F3
InChI KeyJXUKFFRPLNTYIV-UHFFFAOYSA-N
SMILESFC1=CC(F)=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Refractive Index1.4120-1.4160 @ 20?C
1,3,5-Trifluorobenzene was used in the synthesis of (-)-epicatechin and its 3-O-gallate. It is also used as an intermediate in organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trifluorobenzene was used in the synthesis of (-)-epicatechin and its 3-O-gallate. It is also used as an intermediate in organic chemical synthesis.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. H.Z.Grotto; A.C.Vigoritto; J.F.Noronha; G.A.Lima. A new synthetic strategy for catechin-class polyphenols: concise synthesis of (-)-epicatechin and its 3-O-gallate. Chemical Communications. 2012, 48, (67),8425-8427
  2. T.E.Bull. Extended diffusion of 1,3,5-trifluorobenzene-d 3 and acetonitrile-d 3 by spin-lattice relaxation. J. Chem. Phys. 1975, 62, (1),222