Lithium dodecylsulfate, 98%
Lithium dodecylsulfate, 98%
Lithium dodecylsulfate, 98%
Thermo Scientific Chemicals

Lithium dodecylsulfate, 98%

CAS: 2044-56-6 | C12H25LiO4S | 272.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A16005.22
also known as A16005-22
Price (EUR)
373,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
373,00
Each
Chemical Identifiers
CAS2044-56-6
IUPAC Namelithium(1+) dodecyl sulfate
Molecular FormulaC12H25LiO4S
InChI KeyYFVGRULMIQXYNE-UHFFFAOYSA-M
SMILES[Li+].CCCCCCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (ex Sulphated Ash)> 97.5%
Sulfated Ash19.68 - 20.69%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only