N-(n-Butyl)phthalimide, 99%
N-(n-Butyl)phthalimide, 99%
N-(n-Butyl)phthalimide, 99%
N-(n-Butyl)phthalimide, 99%
Thermo Scientific Chemicals

N-(n-Butyl)phthalimide, 99%

CAS: 1515-72-6 | C12H13NO2 | 203.241 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16020.06
also known as A16020-06
Price (EUR)
29,20
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
29,20
Each
Add to cart
Chemical Identifiers
CAS1515-72-6
IUPAC Name2-butyl-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC12H13NO2
InChI KeyDLKDEVCJRCPTLN-UHFFFAOYSA-N
SMILESCCCCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥98.5%
Melting Point (clear melt)28.5-34.5?C
FormCrystals or powder or crystalline powder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only