2-(2-n-Butoxyethoxy)ethyl acetate, 98%
2-(2-n-Butoxyethoxy)ethyl acetate, 98%
2-(2-n-Butoxyethoxy)ethyl acetate, 98%
Thermo Scientific Chemicals

2-(2-n-Butoxyethoxy)ethyl acetate, 98%

CAS: 124-17-4 | C10H20O4 | 204.266 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1000 mL
Catalog number A16065.AU
also known as A16065-AU
Price (EUR)
44,00
Each
Quantity:
1000 mL
Request bulk or custom format
Price (EUR)
44,00
Each
Chemical Identifiers
CAS124-17-4
IUPAC Name2-(2-butoxyethoxy)ethyl acetate
Molecular FormulaC10H20O4
InChI KeyVXQBJTKSVGFQOL-UHFFFAOYSA-N
SMILESCCCCOCCOCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4235-1.4275 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
2-(2-n-Butoxyethoxy)ethyl acetate is widely used in high-grade paints for car, TV, freezer and plane. It is used as a solvent for enhancement of coating's characters in the coating industry. It is also used as a cleaning and furnishing care product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-n-Butoxyethoxy)ethyl acetate is widely used in high-grade paints for car, TV, freezer and plane. It is used as a solvent for enhancement of coating′s characters in the coating industry. It is also used as a cleaning and furnishing care product.

Solubility
Miscible with water, acetone, ethyl ether, ethanol and most organic liquids.

Notes
Light sensitive and heat sensitive. Incompatible with strong oxidizing agents, acids and strong bases. Keep the container tightly closed.
RUO – Research Use Only

General References:

  1. Zhou, J.; Gan, W.; Tang, H.; Li, Y.; Yang, C. One-pot synthesis of Ag nanoparticle-coated Pb-based glass frit used in crystalline silicon solar cell. Appl. Phys. A 2015, 118 (4), 1371-1376.
  2. Khan, A.; Khan, A. A. P.; Rahman, M. M.; Asiri, A. M.; Al-Youbi, A. O. Toward designing efficient rice-shaped polyaniline@bismuth oxide nanocomposites for sensor application. J. Sol-Gel Sci. Technol. 2015, 76 (3), 519-528.