Methyl 2-fluorobenzoate, 98%
Methyl 2-fluorobenzoate, 98%
Methyl 2-fluorobenzoate, 98%
Methyl 2-fluorobenzoate, 98%
Thermo Scientific Chemicals

Methyl 2-fluorobenzoate, 98%

CAS: 394-35-4 | C8H7FO2 | 154.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16133.06
also known as A16133-06
Price (EUR)
13,50
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
13,50
Each
Add to cart
Chemical Identifiers
CAS394-35-4
IUPAC Namemethyl 2-fluorobenzoate
Molecular FormulaC8H7FO2
InChI KeyQAFJIJWLEBLXHH-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
Refractive Index1.5000-1.5040 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only