Copper, powder, 99+%
Copper, powder, 99+%
Copper, powder, 99+%
Thermo Scientific Chemicals

Copper, powder, 99+%

CAS: 7440-50-8 | Cu
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A16243.0E
also known as A16243-0E
Price (EUR)
-
Quantity:
2500 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point2595°C
CAS7440-50-8
Melting Point1083°C
Molecular FormulaCu
Percent Purity≥99%
Quantity2500 g
Unit SizeEach