Diiodofluorescein
Diiodofluorescein
Diiodofluorescein
Thermo Scientific Chemicals

Diiodofluorescein

CAS: 31395-16-1 | C20H10I2O5 | 584.104 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A16554.18
also known as A16554-18
Price (EUR)
124,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
124,00
Each
Chemical Identifiers
CAS31395-16-1
IUPAC Name3',6'-dihydroxy-4,5-diiodo-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular FormulaC20H10I2O5
InChI KeyRUEQYKGDINYYHI-UHFFFAOYSA-N
SMILESOC1=CC2=C(C=C1)C1(OC(=O)C3=C1C=CC(I)=C3I)C1=C(O2)C=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red to brown
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only