2'-Nitroacetanilide, 98+%
2'-Nitroacetanilide, 98+%
2'-Nitroacetanilide, 98+%
2'-Nitroacetanilide, 98+%
Thermo Scientific Chemicals

2'-Nitroacetanilide, 98+%

CAS: 552-32-9 | C8H8N2O3 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16677.06
also known as A16677-06
Price (EUR)
25,16
Online Exclusive
29,60
Save 4,44 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
25,16
Online Exclusive
29,60
Save 4,44 (15%)
Each
Add to cart
Chemical Identifiers
CAS552-32-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Melting Point (clear melt)88.0-94.0?C
Assay (GC)≥98.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only