3-Benzoylpyridine, 98+%
3-Benzoylpyridine, 98+%
3-Benzoylpyridine, 98+%
Thermo Scientific Chemicals

3-Benzoylpyridine, 98+%

CAS: 5424-19-1 | C12H9NO | 183.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A16695.06
Price (EUR)
36,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
36,30
Each
Chemical Identifiers
CAS5424-19-1
IUPAC Name3-benzoylpyridine
Molecular FormulaC12H9NO
InChI KeyRYMBAPVTUHZCNF-UHFFFAOYSA-N
SMILESO=C(C1=CC=CC=C1)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused solid or clear liquid as melt
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Appearance (Color)Pale cream to cream to yellow
Melting Point (clear melt)34.0-41.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only