8-Aminoquinoline, 98+%
8-Aminoquinoline, 98+%
8-Aminoquinoline, 98+%
Thermo Scientific Chemicals

8-Aminoquinoline, 98+%

CAS: 578-66-5 | C9H8N2 | 144.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A16752.03
also known as A16752-03
Price (EUR)
35,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
35,60
Each
Chemical Identifiers
CAS578-66-5
IUPAC Namequinolin-8-amine
Molecular FormulaC9H8N2
InChI KeyWREVVZMUNPAPOV-UHFFFAOYSA-N
SMILESNC1=C2N=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)62.0-68.0?C
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Appearance (Color)Cream to yellow to brown
FormCrystals or powder or crystalline powder or liquid as melt
8-Aminoquinoline has been used in the preparation of base-stabilized terminal borylene complex of osmium. It is also used in the spectrophotometric determination of bivalent palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Aminoquinoline has been used in the preparation of base-stabilized terminal borylene complex of osmium. It is also used in the spectrophotometric determination of bivalent palladium.

Solubility
Soluble in water.

Notes
Air Sensitive. Store away from air under dry inert gas. Incompatible with air and oxidizing agents.
RUO – Research Use Only

General References:

  1. S G Schulman; L B Sanders. Fluorescence and phosphorescence of 5- and 8-aminoquinoline. Analytica Chimica Acta.1971, 56 (1), 83-89.
  2. Xiaowei Cao; Weiying Lin; Longwei He. A near-infrared fluorescence turn-on sensor for sulfide anions. Organic Letters. 2011, 13 (17), 4716-4719.