Naphthalene-2-sulfonyl chloride, 97%
Naphthalene-2-sulfonyl chloride, 97%
Naphthalene-2-sulfonyl chloride, 97%
Naphthalene-2-sulfonyl chloride, 97%
Thermo Scientific Chemicals

Naphthalene-2-sulfonyl chloride, 97%

CAS: 93-11-8 | C10H7ClO2S | 226.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A16872.36
also known as A16872-36
Price (EUR)
651,00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
651,00
Each
Add to cart
Chemical Identifiers
CAS93-11-8
IUPAC Namenaphthalene-2-sulfonyl chloride
Molecular FormulaC10H7ClO2S
InChI KeyOPECTNGATDYLSS-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow to pale brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Assay (GC)≥96.0%
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only