8-Methoxypsoralen, 98%
8-Methoxypsoralen, 98%
8-Methoxypsoralen, 98%
8-Methoxypsoralen, 98%
Thermo Scientific Chemicals

8-Methoxypsoralen, 98%

CAS: 298-81-7 | C12H8O4 | 216.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A17108.03
also known as A17108-03
Price (EUR)
61,20
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
61,20
Each
Add to cart
Chemical Identifiers
CAS298-81-7
IUPAC Name9-methoxy-7H-furo[3,2-g]chromen-7-one
Molecular FormulaC12H8O4
InChI KeyQXKHYNVANLEOEG-UHFFFAOYSA-N
SMILESCOC1=C2OC=CC2=CC2=C1OC(=O)C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥97.5% (NMR) (U.S. specification)
CommentPurchased in U.K. and U.S.
Melting Point143-148°C (U.K. specification)
Proton NMRConforms to structure (U.S. specification)
Appearance (Color)White to pale cream or pale yellow
View more
8-Methoxypsoralen, is used in Photochemotherapy (methoxsalen with long wave ultraviolet radiation) is indicated for the repigmentation of idiopathic vitiligo. It is also used in Photopheresis (methoxsalen with long wave ultraviolet radiation of white blood cells) is indicated for use with the UVAR* System in the palliative treatment of the skin manifestations of cutaneous T-cell lymphoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Methoxypsoralen, is used in Photochemotherapy (methoxsalen with long wave ultraviolet radiation) is indicated for the repigmentation of idiopathic vitiligo. It is also used in Photopheresis (methoxsalen with long wave ultraviolet radiation of white blood cells) is indicated for use with the UVAR* System in the palliative treatment of the skin manifestations of cutaneous T-cell lymphoma.

Solubility
Soluble in acetone, ethanol (slight), acetic acid, and chloroform (50 mg/ml). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. NJ Lowe.; D Weingarten.; T Bourget, LS Moy. PUVA therapy for psoriasis: comparison of oral and bath-water delivery of 8-methoxypsoralen. Journal of the American Academy of Dermatology. 198614 (5), 754-760.
  2. S Igali.; BA Bridges.; MJ Ashwood-Smith. Mutagenesis in Escherichia coli IV. Photosensitization to near ultraviolet light by 8-methoxypsoralen. Journal of the American Academy of Dermatology. 19709 (1), 21-30.