Lithium formate hydrate, 98%
Lithium formate hydrate, 98%
Lithium formate hydrate, 98%
Thermo Scientific Chemicals

Lithium formate hydrate, 98%

CAS: 6108-23-2 | CHLiO2 | 51.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A17432.18
also known as A17432-18
Price (EUR)
29,70
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
29,70
Each
Chemical Identifiers
CAS6108-23-2
IUPAC Namelithium(1+) formate
Molecular FormulaCHLiO2
InChI KeyXKPJKVVZOOEMPK-UHFFFAOYSA-M
SMILES[Li+].[O-]C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5% (Dry wt. basis)
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)14.77-34.21% (0.5-1.5 waters)
Appearance (Color)White
It is used in the investigation of signal fading in lithium formate EPR dosimeters using a new sensitive method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the investigation of signal fading in lithium formate EPR dosimeters using a new sensitive method.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. S. Lelie, et al. The investigation of lithium formate hydrate, sodium dithionate and N-methyl taurine as clinical EPR dosimeters.Radiation Measurements,2013,59218-224.
  2. Y. Epelboin, et al. Determination of Burgers vectors of dislocations in synthetic quartz by computer simulation.J. Appl. Phys.,1982,53571.