Magnesium trifluoromethanesulfonate, 97%
Magnesium trifluoromethanesulfonate, 97%
Magnesium trifluoromethanesulfonate, 97%
Magnesium trifluoromethanesulfonate, 97%
Thermo Scientific Chemicals

Magnesium trifluoromethanesulfonate, 97%

CAS: 60871-83-2 | C2F6MgO6S2 | 322.431 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A17591.06
also known as A17591-06
Price (EUR)
35,50
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
35,50
Each
Add to cart
Chemical Identifiers
CAS60871-83-2
IUPAC Namemagnesium(2+) ditrifluoromethanesulfonate
Molecular FormulaC2F6MgO6S2
InChI KeyBZQRBEVTLZHKEA-UHFFFAOYSA-L
SMILES[Mg++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay≥96.0% to ≤104.0%
Appearance (Color)White to very pale cream
FormPowder
Loss on Drying≤3.5% (105°C/constant wt.)
Magnesium trifluoromethanesulfonate is used as a component of the electrolyte, as lewis acid resulted in largely decreased and reversed enantioselectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium trifluoromethanesulfonate is used as a component of the electrolyte, as lewis acid resulted in largely decreased and reversed enantioselectivities.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. G.P. Pandey.; S.A. Hashmi. Experimental investigations of an ionic-liquid-based, magnesium ion conducting, polymer gel electrolyte. Journal of Power Sources. 2009, 187, (2), 627-634.
  2. J.L. Acosta.; E. Morales. Synthesis and characterization of polymeric electrolytes for solid state magnesium batteries. Electrochimica Acta. 1998, 43, (7), 791-797.