3,6-Dithia-1,8-octanediol, 97%
3,6-Dithia-1,8-octanediol, 97%
3,6-Dithia-1,8-octanediol, 97%
Thermo Scientific Chemicals

3,6-Dithia-1,8-octanediol, 97%

CAS: 5244-34-8 | C6H14O2S2 | 182.296 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A17654.09
also known as A17654-09
Price (EUR)
23,90
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
23,90
Each
Specifications
Beilstein1739193
CAS5244-34-8
Chemical Name or Material3,6-Dithia-1,8-octanediol
EINECS Number226-045-8
Formula Weight182.31
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only