Benzoylferrocene, 98%, Thermo Scientific Chemicals
Benzoylferrocene, 98%, Thermo Scientific Chemicals
Benzoylferrocene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzoylferrocene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A17685.03
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1272-44-2
IUPAC NameBenzoylferrocene
Molecular FormulaC17H14FeO
SMILES[Fe].c1cccc1.O=C(c1cccc1)C1=CC=CC=C1
Molecular Weight (g/mol)290.14
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Red to brown
Assay (GC)≥97.5%
Melting Point (clear melt)104.0-110.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only