4-Amino-1,2,4-triazole, 99%
4-Amino-1,2,4-triazole, 99%
4-Amino-1,2,4-triazole, 99%
Thermo Scientific Chemicals

4-Amino-1,2,4-triazole, 99%

CAS: 584-13-4 | C2H4N4 | 84.082 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A17716.06
also known as A17716-06
Price (EUR)
28,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
28,00
Each
Chemical Identifiers
CAS584-13-4
IUPAC Name4H-1,2,4-triazol-4-amine
Molecular FormulaC2H4N4
InChI KeyFMCUPJKTGNBGEC-UHFFFAOYSA-N
SMILESNN1C=NN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)≤0.75%
Appearance (Color)White
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder or fused solid
View more
4-Amino-1,2,4-triazole is used as an intermediate for the preparation of antifungal agent like fluconazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1,2,4-triazole is used as an intermediate for the preparation of antifungal agent like fluconazole.

Solubility
Soluble in water and methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Berner, U.; Widenmeyer, M. W.; Engler, P.; Dale, P. J. 4-Amino-1,2,4-triazole: Playing a key role in the chemical deposition of Cu-In-Ga metal layers for photovoltaic applications. Thin Solid Films 2015, 582, 39-42.
  2. Xu, C. X.; Zhang, J. G.; Yin, X.; Jin, X.; Li, T.; Zhang, T. L.; Zhou, Z. N. Cd(II) complexes with different nuclearity and dimensionality based on 3-hydrazino-4-amino-1,2,4-triazole. J. Solid State Chem. 2015, 226, 59-65.