Sodium formate, 98%
Sodium formate, 98%
Sodium formate, 98%
Thermo Scientific Chemicals

Sodium formate, 98%

CAS: 141-53-7 | CHNaO2 | 68.007 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A17813.0I
Price (EUR)
110,65
線上優惠
123,00
Save 12,35 (10%)
Each
Quantity:
5000 g
Request bulk or custom format
Price (EUR)
110,65
線上優惠
123,00
Save 12,35 (10%)
Each
Chemical Identifiers
CAS141-53-7
IUPAC Namesodium formate
Molecular FormulaCHNaO2
InChI KeyHLBBKKJFGFRGMU-UHFFFAOYSA-M
SMILES[Na+].[O-]C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or pale cream
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1.0%
Sodium formate is used in analytical chemistry to precipitate noble metals. It is used for tanning leather, for dyeing and printing fabrics. It acts as an electroplating agent, an acidulant in the textile industry, a reducing agent, a mordant, a complexing agent. Further, it is used in photographic fixing baths. In addition, it is used as a buffering agent for strong mineral acids and food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium formate is used in analytical chemistry to precipitate noble metals. It is used for tanning leather, for dyeing and printing fabrics. It acts as an electroplating agent, an acidulant in the textile industry, a reducing agent, a mordant, a complexing agent. Further, it is used in photographic fixing baths. In addition, it is used as a buffering agent for strong mineral acids and food additive.

Solubility
Soluble in water, alcohol, glycerol and formic acid. Insoluble in ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids. Moisture sensitive
RUO – Research Use Only

General References:

  1. Converts Grignard reagents to aldehydes in excellent yields: Tetrahedron Lett., 25, 1843 (1984). Lithium formate may also be used.
  2. Under phase-transfer conditions converts primary alkyl chlorides to alcohols via the formate esters: Synthesis, 763 (1986).
  3. Reaction with acetyl chloride gives the useful formylating agent, formic acetic anhydride. For details, including isolation by distillation, see: Org. Synth. Coll., 6, 8 (1988); review: Tetrahedron, 46, 1081 (1990).
  4. Reduces amine oxides to amines under mild conditions: Chem. Lett., 1517 (1985).
  5. Can be used as a hydrogen donor in catalytic transfer hydrogenation reactions. More active hydrogen donor than formic acid in hydrogenolysis of aryl chlorides. For a comparative mechanistic study of formic acid and formate salts in this reaction, see: J. Org. Chem., 60, 1347 (1995). See also Ammonium formate, A10699 .
  6. Kathó, Á.; Szatmári, I.; Papp, G.; Joó, F. Effect of 2-Propanol on the Transfer Hydrogenation of Aldehydes by Aqueous Sodium Formate using a Rhodium(I)-sulfonated Triphenylphosphine Catalyst. Int. J. Chem. 2015, 69 (6), 339-344.
  7. Soni, R.; Hall, T. H.; Mitchell, B. P.; Owen, M. R.; Wills, M. Asymmetric reduction of electron-rich ketones with tethered ru(II)/TsDPEN catalysts using formic acid/triethylamine or aqueous sodium formate. J. Org. Chem. 2015, 80 (13), 6784-6793.