2-Amino-2-methyl-1-propanol, 95%, may cont. ca 5% water
2-Amino-2-methyl-1-propanol, 95%, may cont. ca 5% water
2-Amino-2-methyl-1-propanol, 95%, may cont. ca 5% water
Thermo Scientific Chemicals

2-Amino-2-methyl-1-propanol, 95%, may cont. ca 5% water

CAS: 124-68-5 | C4H11NO | 89.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number A17814.AE
also known as A17814-AE
Price (EUR)
26,40
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
26,40
Each
Chemical Identifiers
CAS124-68-5
IUPAC Name2-amino-2-methylpropan-1-ol
Molecular FormulaC4H11NO
InChI KeyCBTVGIZVANVGBH-UHFFFAOYSA-N
SMILESCC(C)(N)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4420-1.4485 @ 20?C
Appearance (Color)Clear colorless
FormViscous liquid or low melting solid
Assay (GC)≥94.0%
Water Content (Karl Fischer Titration)<6.0%
View more
2-Amino-2-methyl-1-propanol is used for the preparation of buffer solution and in cosmetics. It is also used in ATR-FTIR spectroscopic investigation of the carbon monoxide absorption characteristics of a series of heterocyclic diamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-2-methyl-1-propanol is used for the preparation of buffer solution and in cosmetics. It is also used in ATR-FTIR spectroscopic investigation of the carbon monoxide absorption characteristics of a series of heterocyclic diamines.

Solubility
Immiscible with water. Miscible with alcohols.

Notes
Incompatible withoxidizing agents, strong acids, copper, brass and aluminum.
RUO – Research Use Only

General References:

  1. Buffer and phosphate acceptor in assay of phosphatases: Methods of Enzymatic Analysis, 3rd ed., H. U. Bergmeyer, Ed., Verlag Chemie, Weinheim (1984), vol. 4, p76.
  2. Precursor of 2-substituted-4,4-dimethyl-2-oxazoline derivatives of carboxylic acids: J. Org. Chem., 39, 2787 (1974), developed by Meyers. The oxazoline, which is readily formed with the acid chloride and thionyl chloride, is stable to base, organometallic reagents, etc., but readily cleaved by dilute acid. For further information on the reactivity of these derivatives, see 2,4,4-Trimethyl-2-oxazoline, L00181.
  3. 2-Aryl-4,4-dimethyl-2-oxazolines are activated towards ortho-lithiation: J. Org. Chem., 40, 3058 (1975). Methoxy- or fluoro-substituents in the ortho-position are readily substitutied by organolithiums or Grignards, providing a versatile route to biaryls: J. Am. Chem. Soc ., 97, 7383 (1975). For examples, see: Org. Synth. Coll., 9, 258 (1998). The oxazoline can also be converted to an aldehyde by methylation and borohydride reduction.
  4. For reviews of the use of oxazolines in synthesis, see: Angew. Chem. Int. Ed., 15, 270 (1976); Tetrahedron, 41, 837 (1985); 50, 2297 (1994)./
  5. Conway, W.; Bruggink, S.; Beyad, Y.; Luo, W.; Cabrera, I. M., Puxty, G.; Feron, P. CO2 absorption into aqueous amine blended solutions containing monoethanolamine (MEA), N, N-dimethylethanolamine (DMEA), N, N-diethylethanolamine (DEEA) and 2-amino-2-methyl-1-propanol (AMP) for post-combustion capture processes. Chem. Eng. Sci. 2015, 126, 446-454.
  6. Halim, H. N. A.; Shariff, A. M.; Bustam, M. A. High pressure CO2 absorption from natural gas using piperazine promoted 2-amino-2-methyl-1-propanol in a packed absorption column. Sep. Purif. Technol. 2015, 152, 87-93.