1,2,4,5-Tetrafluorobenzene, 99%
1,2,4,5-Tetrafluorobenzene, 99%
1,2,4,5-Tetrafluorobenzene, 99%
Thermo Scientific Chemicals

1,2,4,5-Tetrafluorobenzene, 99%

CAS: 327-54-8 | C6H2F4 | 150.076 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17815.06
also known as A17815-06
Price (EUR)
68,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
68,20
Each
Chemical Identifiers
CAS327-54-8
IUPAC Name1,2,4,5-tetrafluorobenzene
Molecular FormulaC6H2F4
InChI KeySDXUIOOHCIQXRP-UHFFFAOYSA-N
SMILESFC1=CC(F)=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4055-1.4085 @ 20?C
FormLiquid
It is employed as intermediate in organic synthesis. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate in organic synthesis. It is also used as pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources.
RUO – Research Use Only

General References:

  1. Katsuhiko Okuyama.; Takeo Kakinuma.; Masaaki Fujii.; Naohiko Mikami.; Mitsuo Ito. Electronic spectra of 1,2,4,5-tetrafluorobenzene in a supersonic jet: butterfly tunneling in the excited state.J. Phys. Chem.,1986, 90(17), 3948-3952.
  2. G. C. Finger.; F. H. Reed.; D. M. Burness.; D. M. Fort.; R. R. Blough. Aromatic Fluorine Compounds. II. 1,2,4,5-Tetrafluorobenzene and Related Compounds1,2.J. Am. Chem. Soc. 1951, 73(1), 145-149.