Manganese(III) phosphate hydrate, 99%
Manganese(III) phosphate hydrate, 99%
Manganese(III) phosphate hydrate, 99%
Thermo Scientific Chemicals

Manganese(III) phosphate hydrate, 99%

CAS: 104663-56-1 | MnO4P | 149.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A17868.18
also known as A17868-18
Price (EUR)
84,90
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
84,90
Each
Chemical Identifiers
CAS104663-56-1
IUPAC Namemanganese(3+) phosphate
Molecular FormulaMnO4P
InChI KeyBECVLEVEVXAFSH-UHFFFAOYSA-K
SMILES[Mn+3].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale grey to pale brown or green
FormPowder or crystalline powder
Complexometric Titration(Manganese, Mn): 36.1-37.2% (dry wt. basis)
Complexometric Titration≥98.5 to ≤101.5% (dry wt. basis)
Loss on Drying≤20% (200°C/constant wt.)
It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Insoluble in water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. F. W. Boyle; and W. L. Lindsay. Preparation, X-Ray Diffraction Pattern, and Solubility Product of Manganese(III) Phosphate Hydrate. Soil Science Society of America Journal, 1984,49(3), 758-760.
  2. S.M.S. Chauhan; B.B. Sahoo. Biomimetic oxidation of ibuprofen with hydrogen peroxide catalysed by horseradish peroxidase (HRP) and 5,10,15,20- tetrakis-(2',6'-dichloro-3'-sulphonatophenyl)porphyrinatoiron(III) and manganese(III) hydrates in AOT reverse micelles. Bioorganic & Medicinal Chemistry, 1999,7(11), 2629-2634.