4-Bromostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
4-Bromostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
4-Bromostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
4-Bromostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
Thermo Scientific Chemicals

4-Bromostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

CAS: 2039-82-9 | C8H7Br | 183.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A17896.22
also known as A17896-22
Price (EUR)
582,25
Online Exclusive
685,00
Save 102,75 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
582,25
Online Exclusive
685,00
Save 102,75 (15%)
Each
Add to cart
Chemical Identifiers
CAS2039-82-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.5915-1.5965 @ 20°C
Solution Test(10% v/v in diethyl ether) : Clear
Solution Test(10% v/v in methanol) : Clear
Appearance (Color)Clear colorless to yellow
View more
4-Bromostyrene is widely utilized in the synthesis of nitroolefins by alkene cross-metathesis. It plays an important role in the Heck reaction to the synthesis of poly(1,4-phenylenevinylene). It is employed in the structure activity relationships (SAR) study of the chemical and biochemical properties of the vinyl group of styrene. It is also used to investigate the photochemical growth of Br-terminated self-assembled monolayers. It is also involved in the synthesis of silsesquioxanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromostyrene is widely utilized in the synthesis of nitroolefins by alkene cross-metathesis. It plays an important role in the Heck reaction to the synthesis of poly(1,4-phenylenevinylene). It is employed in the structure activity relationships (SAR) study of the chemical and biochemical properties of the vinyl group of styrene. It is also used to investigate the photochemical growth of Br-terminated self-assembled monolayers. It is also involved in the synthesis of silsesquioxanes.

Solubility
Miscible with ethanol, ether and benzene.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, bases, oxidizing agents and halogens. Keep away from sources of ignition.
RUO – Research Use Only

General References:

  1. Zhang, Y. P.; He, J. H.; Xu, G. Q. Selective Attachment of 4-Bromostyrene on the Si(111)-(7 x 7) Surface. J. Phys. Chem. C 2011, 115 (31), 15496-15501.
  2. Demirel, G. B.; Çakmak, M.; Çaykara, T. Understanding 4-bromostyrene Adsorption on the Si(001)-(1 x 2) surface: A density functional theory study. Surf. Sci. 2011, 605 (11-12), 1056-1061.