2,2-Diphenylpropionitrile, 97%
2,2-Diphenylpropionitrile, 97%
2,2-Diphenylpropionitrile, 97%
Thermo Scientific Chemicals

2,2-Diphenylpropionitrile, 97%

CAS: 5558-67-8 | C15H13N | 207.276 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Catalog number A17991.09
also known as A17991-09
Price (EUR)
38,50
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
38,50
Each
Chemical Identifiers
CAS5558-67-8
IUPAC Name2,2-diphenylpropanenitrile
Molecular FormulaC15H13N
InChI KeyDPVHBXFSKLKYIQ-UHFFFAOYSA-N
SMILESCC(C#N)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid or viscous liquid
Assay (GC)≥96.0%
Refractive Index1.5710-1.5760 @ 20?C
Appearance (Color)Clear yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only