2,6-Difluorophenylacetic acid, 98%
2,6-Difluorophenylacetic acid, 98%
2,6-Difluorophenylacetic acid, 98%
Thermo Scientific Chemicals

2,6-Difluorophenylacetic acid, 98%

CAS: 85068-28-6 | C8H6F2O2 | 172.131 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18063.14
also known as A18063-14
Price (EUR)
211,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
211,00
Each
Chemical Identifiers
CAS85068-28-6
IUPAC Name2-(2,6-difluorophenyl)acetic acid
Molecular FormulaC8H6F2O2
InChI KeyFUGDCKXBUZFEON-UHFFFAOYSA-N
SMILESOC(=O)CC1=C(F)C=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Melting Point (clear melt)97.0-103.0?C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only