Nigrosin water soluble
Nigrosin water soluble
Nigrosin water soluble
Thermo Scientific Chemicals

Nigrosin water soluble

CAS: 8005-03-6 | C22H14N6Na2O9S2 | 616.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A18147.14
also known as A18147-14
Price (EUR)
22,20
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
22,20
Each
Chemical Identifiers
CAS8005-03-6
IUPAC Namedisodium (3E)-5-amino-6-[(1E)-2-(4-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Molecular FormulaC22H14N6Na2O9S2
InChI KeyHKBVRFLHNUEVRO-HMDKGIQMSA-L
SMILES[Na+].[Na+].NC1=C2C(=O)\C(=N/NC3=CC=CC=C3)C(=CC2=CC(=C1\N=N\C1=CC=C(C=C1)[N+]([O-])=O)S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
lambda-max (UV-Vis)560-590nm (water)
Appearance (Color)Black
Specific Absorbance (E 1%/1cm)>200 at Lambda max (water)
Loss on Drying≤15% (105?C, constant weight)
View more
It is employed as a common biological stain for tissues and cells. Used to make inks and shoe-polish and for dyeing leather, wood, textiles, and plastics. The crude mixture of nigrosine hydrochlorides (Phenol Black L) is used to dye phenolic resins. Bases are used to make printing colors and to dye plastics. It finds its use in Dorner?s spore stain, as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi. It is also used in compounding resins include phenolic resins, styrenics, polyamides and urea resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a common biological stain for tissues and cells. Used to make inks and shoe-polish and for dyeing leather, wood, textiles, and plastics. The crude mixture of nigrosine hydrochlorides (Phenol Black L) is used to dye phenolic resins. Bases are used to make printing colors and to dye plastics. It finds its use in Dorner′s spore stain, as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi. It is also used in compounding resins include phenolic resins, styrenics, polyamides and urea resins.

Solubility
Soluble in water, ethanol, oil, nigrosines and fat.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Zhang S.; Zhao XW.; Xu H.; Zhu R.; Gu ZZ. Fabrication of photonic crystals with nigrosine-doped poly(MMA-co-DVB-co-MAA) particles.J Colloid Interface Sci.2007,316(1), 168-74.
  2. J.P. Kaltenbach.; Merle H. Kaltenbach.; W.B. Lyons. Nigrosin as a dye for differentiating live and dead ascites cells.Experimental Cell Research,1958,15(1), 112-117.