4-tert-Butyltoluene, 95%
4-tert-Butyltoluene, 95%
4-tert-Butyltoluene, 95%
Thermo Scientific Chemicals

4-tert-Butyltoluene, 95%

CAS: 98-51-1 | C11H16 | 148.249 g/mol
Have Questions?
Catalog NumberQuantity
A18214.AP
also known as A18214-AP
500 mL
Catalog number A18214.AP
also known as A18214-AP
Price (EUR)
43,60
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
43,60
Each
Chemical Identifiers
CAS98-51-1
IUPAC Name1-tert-butyl-4-methylbenzene
Molecular FormulaC11H16
InChI KeyQCWXDVFBZVHKLV-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥94.0%
Identification (FTIR)Conforms
Refractive Index1.4890-1.4950 @ 20?C
FormLiquid
4-tert-Butyltoluene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butyltoluene is used as pharmaceutical intermediate.

Solubility
Soluble in water (6 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Shivanand Pai; Upendranath Gupta; Satyanarayana Chilukuri. Butylation of toluene: Influence of zeolite structure and acidity on 4-tert-butyltoluene selectivity. Journal of Molecular Catalysis A: Chemical. 2007, 265,(1-2), 109-116.
  2. Ahmed A.Amin; James K.Beattie. Inhibition of the cobalt acetate/bromide-catalyzed hydrogen peroxide oxidation of 4-tert-butyltoluene. Org. Proc. Res. Dev. 2003, 7,(6), 879-882.