n-Butyltin hydroxide oxide, 95%
n-Butyltin hydroxide oxide, 95%
n-Butyltin hydroxide oxide, 95%
Thermo Scientific Chemicals

n-Butyltin hydroxide oxide, 95%

CAS: 2273-43-0 | C4H10O2Sn | 208.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A18290.22
also known as A18290-22
Price (EUR)
68,60
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
68,60
Each
Chemical Identifiers
CAS2273-43-0
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying<3.0% (105?C/ constant wt.)
Appearance (Color)White
CommentMay contain trace of black particulates from manufacturing process
FormPowder
Assay (unspecified)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only