4-Ethoxyphenyl isothiocyanate, 98%
4-Ethoxyphenyl isothiocyanate, 98%
4-Ethoxyphenyl isothiocyanate, 98%
Thermo Scientific Chemicals

4-Ethoxyphenyl isothiocyanate, 98%

CAS: 3460-49-9 | C9H9NOS | 179.237 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18902.06
also known as A18902-06
Price (EUR)
73,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
73,30
Each
Chemical Identifiers
CAS3460-49-9
IUPAC Name1-ethoxy-4-isothiocyanatobenzene
Molecular FormulaC9H9NOS
InChI KeyREEKOMRZYJXXNR-UHFFFAOYSA-N
SMILESCCOC1=CC=C(C=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow
Assay (GC)≥97.5%
FormFused solid
Identification (FTIR)Conforms
Melting Point (clear melt)56.0-63.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only