Calcium formate, 98%
Calcium formate, 98%
Calcium formate, 98%
Thermo Scientific Chemicals

Calcium formate, 98%

CAS: 544-17-2 | C2H2CaO4 | 130.112 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A18916.22
also known as A18916-22
Price (EUR)
39,30
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
39,30
Each
Chemical Identifiers
CAS544-17-2
IUPAC Namecalcium diformate
Molecular FormulaC2H2CaO4
InChI KeyCBOCVOKPQGJKKJ-UHFFFAOYSA-L
SMILES[Ca++].[O-]C=O.[O-]C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Complexometric Titration≥97.5 to ≤102.5%
An industrial chemical compound and faster setting, early strength gain accelerating admixture for concrete or mortars. It is used in the building industry, in the production of feedstuff mixtures, for silage and in the tannery. Also used in leather tanning, producing formic acid, concrete cure accelerator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An industrial chemical compound and faster setting, early strength gain accelerating admixture for concrete or mortars. It is used in the building industry, in the production of feedstuff mixtures, for silage and in the tannery. Also used in leather tanning, producing formic acid, concrete cure accelerator.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Michael Weber; Christine Hellriegel; Alexander Rueck; Juerg Wuethrich; Peter Jenks; Markus Obkircher. Method development in quantitative NMR towards metrologically traceable organic certified reference materials used as (31)P qNMR standards. Analytical and Bioanalytical Chemistry., 2015, 407 (11), 3115-3123.
  2. Michael Weber et. al. Using high-performance1H NMR (HP-qNMR™) for the certification of organic reference materials under accreditation guidelines—Describing the overall process with focus on homogeneity and stability assessment. Journal of Pharmaceutical and Biomedical Analysis, 2014, 93102-110.