Isobutyl isothiocyanate, 97%
Isobutyl isothiocyanate, 97%
Isobutyl isothiocyanate, 97%
Thermo Scientific Chemicals

Isobutyl isothiocyanate, 97%

CAS: 591-82-2 | C5H9NS | 115.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18932.06
also known as A18932-06
Price (EUR)
35,90
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
35,90
Each
Chemical Identifiers
CAS591-82-2
IUPAC Name1-isothiocyanato-2-methylpropane
Molecular FormulaC5H9NS
InChI KeyNSDDRJXKROCWRZ-UHFFFAOYSA-N
SMILESCC(C)CN=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4955-1.5005 @ 20?C
FormLiquid
It is isothiocyanate compound used for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is isothiocyanate compound used for proteomics research.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
RUO – Research Use Only

General References:

  1. S. Dehshahri; S. Afsharypuor; G. Asghari; and A. Mohagheghzadeh. Determination of volatile glucosinolate degradation products in seed coat, stem and in vitro cultures of Moringa peregrina (Forssk.) Fiori. Res Pharm Sci., 2012, 7 (1), 51-56.
  2. Mahmoud A. Al-Qudah; and Musa H. Abu Zarga. Chemical Composition of Essential Oils from Aerial Parts of Sisymbrium Irio from Jordan. E-Journal of Chemistry, 2010, 7(1), 6-10.