3-Mercapto-1,2-propanediol, 90%
3-Mercapto-1,2-propanediol, 90%
3-Mercapto-1,2-propanediol, 90%
Thermo Scientific Chemicals

3-Mercapto-1,2-propanediol, 90%

CAS: 96-27-5 | C3H8O2S | 108.16 g/mol
Have Questions?
Catalog NumberQuantity
A18948.30
also known as A18948-30
250 g
Catalog number A18948.30
also known as A18948-30
Price (EUR)
205,00
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
205,00
Each
Chemical Identifiers
CAS96-27-5
IUPAC Name3-sulfanylpropane-1,2-diol
Molecular FormulaC3H8O2S
InChI KeyPJUIMOJAAPLTRJ-UHFFFAOYNA-N
SMILESOCC(O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid
Assay (Iodometric Titration)≥88.0%
Appearance (Color)Clear colorless to yellow
It generally used as a size-regulating capping agent for nanocrystals. 1-Thioglycerol is also used in bacteriology experiments to induce the synthesis of porphyrin compounds in aerobically growing Escherichia coli. Also used to study the pH-sensitive photoluminescence of aqueous thiol-capped CdTe nanocrystals. 1-Thioglycerol is used to develop and test thiol functionalized copolymers. 1-Thioglycerol is used as a post-modification agent in the generation of non-standard peptide foldamers.Potential substitute for 2-mercaptoethanol, probe for the study of lymphocyte activation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It generally used as a size-regulating capping agent for nanocrystals. 1-Thioglycerol is also used in bacteriology experiments to induce the synthesis of porphyrin compounds in aerobically growing Escherichia coli. Also used to study the pH-sensitive photoluminescence of aqueous thiol-capped CdTe nanocrystals. 1-Thioglycerol is used to develop and test thiol functionalized copolymers. 1-Thioglycerol is used as a post-modification agent in the generation of non-standard peptide foldamers.Potential substitute for 2-mercaptoethanol, probe for the study of lymphocyte activation.

Solubility
Slightly soluble in water and benzene. Soluble in acetone. Insoluble in ether.

Notes
Air Sensitive and Hygroscopic, store away from water/moisture and air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
RUO – Research Use Only

General References:

  1. G T Javor. Thiol-sensitive genes of Escherichia coli. Journal of Bacteriology., 1989, 170 (7), 5607-5613.
  2. G T Javor; C D Stringer; J Ryu. Thiol-sensitive promoters of Escherichia coli. Journal of Bacteriology, 1988, 170(7), 3291-3293.