(+)-Nootkatone, crystalline, 98+%
(+)-Nootkatone, crystalline, 98+%
(+)-Nootkatone, crystalline, 98+%
Thermo Scientific Chemicals

(+)-Nootkatone, crystalline, 98+%

CAS: 4674-50-4 | C15H22O | 218.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A19166.14
also known as A19166-14
Price (EUR)
640,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
640,00
Each
Chemical Identifiers
CAS4674-50-4
IUPAC Name4,4a-dimethyl-6-(prop-1-en-2-yl)-2,3,4,4a,5,6,7,8-octahydronaphthalen-2-one
Molecular FormulaC15H22O
InChI KeyWTOYNNBCKUYIKC-UHFFFAOYNA-N
SMILESCC1CC(=O)C=C2CCC(CC12C)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystalline powder or powder or crystals or fused solid
Assay (GC)≥98.0%
Optical Rotation+187° ± 6° (c=1 in chloroform)
(+)-Nootkatone, crystalline is used as an effective repellent in mosquitos, bed bugs, lone star ticks and head lice. It is also used as a food additive and flavoring material in beverages. It finds application in fragrances, cosmetics and pharmaceuticals. Further, it is used as a flavor material in beverage, frozen dairy and baked good applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-Nootkatone, crystalline is used as an effective repellent in mosquitos, bed bugs, lone star ticks and head lice. It is also used as a food additive and flavoring material in beverages. It finds application in fragrances, cosmetics and pharmaceuticals. Further, it is used as a flavor material in beverage, frozen dairy and baked good applications.

Solubility
Slightly soluble in ethanol and chloroform. Partly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Schulz, S.; Girhard, M.; Gaßmeyer, S. K.; Jäger, V. D.; Schwarze, D.; Vogel, A.; Urlacher, V. B. Selective Enzymatic Synthesis of the Grapefruit Flavor (+)-Nootkatone. ChemCatChem 2015, 7 (4), 601-604.
  2. Omarini, A. B.; Plagemann, I.; Schimanski, S.; Krings, U.; Berger, R. G. Crosses between monokaryons of Pleurotus sapidus or Pleurotus florida show an improved biotransformation of (+)-valencene to (+)-nootkatone. Bioresour. Technol. 2014, 171, 113-119.