Bromoacetyl bromide, 98%
Bromoacetyl bromide, 98%
Bromoacetyl bromide, 98%
Thermo Scientific Chemicals

Bromoacetyl bromide, 98%

CAS: 598-21-0 | C2H2Br2O | 201.845 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A19580.14
also known as A19580-14
Price (EUR)
31,90
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
31,90
Each
Chemical Identifiers
CAS598-21-0
IUPAC Name2-bromoacetyl bromide
Molecular FormulaC2H2Br2O
InChI KeyLSTRKXWIZZZYAS-UHFFFAOYSA-N
SMILESBrCC(Br)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
Refractive Index1.5450-1.5495 @ 20?C
FormLiquid
Bromoacetyl bromide is widely used in the synthesis of fine chemicals, agrochemicals, dyes and pharmaceuticals. As an acylating agent, it reacts with 10H-phenothiazine to give 10-bromoacetyl-10H-phenothiazine. Similarly, it is also involved in the preparation of bromoactylated starch which is a biodegradable thermoplastic copolymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetyl bromide is widely used in the synthesis of fine chemicals, agrochemicals, dyes and pharmaceuticals. As an acylating agent, it reacts with 10H-phenothiazine to give 10-bromoacetyl-10H-phenothiazine. Similarly, it is also involved in the preparation of bromoactylated starch which is a biodegradable thermoplastic copolymer.

Solubility
Immiscible with water.

Notes
Moisture Sensitive. Store in cool place. Hydrolyses readily. Incompatible with strong oxidizing agents, alcohols and strong bases.
RUO – Research Use Only

General References:

  1. Bayramoglu, G.; Akbulut, A.; Ozalp, V. C.; Arica, M. Y. Immobilized lipase on micro-porous biosilica for enzymatic transesterification of algal oil. Chem. Eng. Res. Des. 2015, 95, 12-21.
  2. Olejniczak, J.; Chan, M.; Almutairi, A. Light-Triggered Intramolecular Cyclization in Poly(lactic-co-glycolic acid)-Based Polymers for Controlled Degradation. Macromolecules 2015, 48 (10), 3166-3172.