Ethyl 3-hexenoate, 98%
Ethyl 3-hexenoate, 98%
Ethyl 3-hexenoate, 98%
Thermo Scientific Chemicals

Ethyl 3-hexenoate, 98%

CAS: 2396-83-0 | C8H14O2 | 142.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A19734.06
also known as A19734-06
Price (EUR)
25,80
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
25,80
Each
Chemical Identifiers
CAS2396-83-0
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4245-1.4275 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only