2,8-Bis(trifluoromethyl)-4-hydroxyquinoline, 97%, Thermo Scientific Chemicals
2,8-Bis(trifluoromethyl)-4-hydroxyquinoline, 97%, Thermo Scientific Chemicals
2,8-Bis(trifluoromethyl)-4-hydroxyquinoline, 97%, Thermo Scientific Chemicals
2,8-Bis(trifluoromethyl)-4-hydroxyquinoline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,8-Bis(trifluoromethyl)-4-hydroxyquinoline, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
A19750.06
also known as A19750-06
5 g
Catalog number A19750.06
also known as A19750-06
Price (EUR)
183,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
183,00
Each
Add to cart
Chemical Identifiers
CAS35853-41-9
IUPAC Name2,8-bis(trifluoromethyl)-1,4-dihydroquinolin-4-one
Molecular FormulaC11H5F6NO
InChI KeyJIWHKBAFGFPZKM-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(=O)C2=CC=CC(=C2N1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)White to yellow to brown or pale grey
FormCrystals or powder or crystalline powder
Melting Point (clear melt)128.0-137.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only