4-n-Pentylbenzenesulfonyl chloride, 97%
4-n-Pentylbenzenesulfonyl chloride, 97%
4-n-Pentylbenzenesulfonyl chloride, 97%
Thermo Scientific Chemicals

4-n-Pentylbenzenesulfonyl chloride, 97%

CAS: 73948-18-2 | C11H15ClO2S | 246.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
Catalog number A19929.88
also known as A19929-88
Price (EUR)
30,00
Each
Quantity:
2.5 g
Request bulk or custom format
Price (EUR)
30,00
Each
Chemical Identifiers
CAS73948-18-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Melting Point (clear melt)37.0-46.0?C
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only